Merck
  • Hepatic uptake of atorvastatin: influence of variability in transporter expression on uptake clearance and drug-drug interactions.
Hepatic uptake of atorvastatin: influence of variability in transporter expression on uptake clearance and drug-drug interactions.
Drug metabolism and disposition: the biological fate of chemicals (2014-05-07)
Anna Vildhede, Maria Karlgren, Elin K Svedberg, Jacek R Wisniewski, Yurong Lai, Agneta Norén, Per Artursson
ZUSAMMENFASSUNG
Differences in the expression and function of the organic anion transporting polypeptide (OATP) transporters contribute to interindividual variability in atorvastatin clearance. However, the importance of the bile acid transporter sodium taurocholate cotransporting polypeptide (NTCP, SLC10A1) in atorvastatin uptake clearance (CLupt) is not yet clarified. To elucidate this issue, we investigated the relative contribution of NTCP, OATP1B1, OATP1B3, and OATP2B1 to atorvastatin CLupt in 12 human liver samples. The impact of inhibition on atorvastatin CLupt was also studied, using inhibitors of different isoform specificities. Expression levels of the four transport proteins were quantified by liquid chromatography tandem mass spectrometry. These data, together with atorvastatin in vitro kinetics, were used to predict the maximal transport activity (MTA) and interindividual differences in CLupt of each transporter in vivo. Subsequently, hepatic uptake impairment on coadministration of five clinically interacting drugs was predicted using in vitro inhibitory potencies. NTCP and OATP protein expression varied 3.7- to 32-fold among the 12 sample donors. The rank order in expression was OATP1B1 > OATP1B3 ≈ NTCP ≈ OATP2B1. NTCP was found to be of minor importance in atorvastatin disposition. Instead, OATP1B1 and OATP1B3 were confirmed as the major atorvastatin uptake transporters. The average contribution to atorvastatin uptake was OATP1B1 > OATP1B3 > OATP2B1 > NTCP, although this rank order varied among individuals. The interindividual differences in transporter expression and CLupt resulted in marked differences in drug-drug interactions due to isoform-specific inhibition. We conclude that this variation should be considered in in vitro to in vivo extrapolations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat -Lösung, BioUltra, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magnesiumsulfat, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Gemfibrozil
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, tablet
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.