Merck
Proteome of tolerance fine-tuning in the human pathogen black yeast Exophiala dermatitidis.
Journal of proteomics (2015-07-21)
Donatella Tesei, Gorji Marzban, Martina Marchetti-Deschmann, Hakim Tafer, Elsa Arcalis, Katja Sterflinger
ZUSAMMENFASSUNG
The black yeast Exophiala dermatitidis is a worldwide distributed agent of primary and secondary diseases in both immunocompromised and healthy humans, with a high prevalence in human-made environments. Since thermo-tolerance has a crucial role in the fungus persistence in man-dominated habitat and in its pathogenicity, three incubation temperatures (37, 45, 1 °C) and two time spans (1 h, 1 week) were selected to simulate different environmental conditions and to investigate the effect of temperature on the proteome of E. dermatitidis CBS 525.76. Using a novel protocol for protein extraction from black yeasts, 2-D DIGE could be applied for characterization of changes in total protein spot abundance among the experimental conditions. A total of 32 variable proteins were identified by mass spectrometry. Data about protein functions, localization and pathways were also obtained. A typical stress response under non-optimal temperature could not be observed at the proteome level, whereas a reduction of the metabolic activity, mostly concerning processes as the general carbon metabolism, was detected after exposure to cold. These results suggest that a fine protein modulation takes place following temperature treatment and a repertoire of stable protein might be at the base of E. dermatitidis adaptation to altered growth conditions. E. dermatitidis is a pathogenic black yeast causing neurotropic infections, systemic and subcutaneous disease in a wide range of hosts, including humans. The discovery of the fungus high prevalence in man-made habitats, including sauna facilities, drinking water and dishwashers, generated concern and raised questions about the infection route. In the present work - which is the first contribution on E. dermatitidis proteome - the effect of different temperature conditions on the fungus protein pattern have been analyzed by using a gel-based approach and the temperature responsive proteins have been identified. The absence of a typical stress response following the exposure to non-optimal temperature was detected at the proteome level, along with a general reduction of the metabolic activity after exposure to cold. These results suggest that a very fine regulation of the protein expression as well as adaptations involving a basic set of stable proteins may be at the base of E. dermatitidis enormous ecological plasticity, which plays a role in the fungus distribution, also enabling the transition from natural to human habitat and to the human host.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Phenol, Molecular Biology