Merck
  • Effect of bioactive glass particles on osteogenic differentiation of adipose-derived mesenchymal stem cells seeded on lactide and caprolactone based scaffolds.
Effect of bioactive glass particles on osteogenic differentiation of adipose-derived mesenchymal stem cells seeded on lactide and caprolactone based scaffolds.
Journal of biomedical materials research. Part A (2015-06-16)
Aitor Larrañaga, Ana Alonso-Varona, Teodoro Palomares, Eva Rubio-Azpeitia, Pablo Aldazabal, Francisco Javier Martin, Jose-Ramon Sarasua
ZUSAMMENFASSUNG
Incorporation of bioactive glass (BG) particles to synthetic polymer scaffolds is a promising strategy to improve the bioactivity of bioinert materials and to stimulate specific cell responses. In this study, the influence of incorporating BG particles to lactide and caprolactone based porous scaffolds on osteogenic differentiation of adipose-derived stem cells (ASCs) was analyzed. Accordingly, ASCs were seeded on poly(L-lactide) (PLLA), poly(ε-caprolactone) (PCL), or poly(L-lactide-co-ε-caprolactone) (PLCL) scaffolds containing 15 vol % of BG particles in two culture conditions: standard versus osteogenic culture medium. In standard culture medium, incorporation of BG to a PLLA scaffold increased the ALP activity with respect to its unfilled counterpart (ca. 1.2- and a 1.6-fold increase over 7 and 14 days, respectively). Moreover, in all the studied polymers the incorporation of BG induced a slightly higher production of mineralized matrix by ASCs, but the differences observed were not statistically significant. In the osteogenic medium, the effect of BG was masked by the effect of osteogenic supplements in the long-term. However, in the short-term (day 7), BG particles induced an early ALP activity of predifferentiated osteoblasts on PLLA and PCL scaffolds and higher matrix mineralization on PCL scaffolds. In summary, the addition of BG particles to PLLA and PCL scaffolds sustains ASC osteogenic differentiation, facilitates mineralization and induces the formation of a hydroxyapatite layer on the surface of the polymer scaffolds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O