Merck
  • A comparative study on effect of metformin and metformin-conjugated nanotubes on blood glucose homeostasis in diabetic rats.
A comparative study on effect of metformin and metformin-conjugated nanotubes on blood glucose homeostasis in diabetic rats.
European journal of drug metabolism and pharmacokinetics (2014-06-28)
Naser Mirazi, Jamileh Shoaei, Ardeshir Khazaei, Abdolkarim Hosseini
ZUSAMMENFASSUNG
Diabetes mellitus is one of the most prevalent metabolic disorders. Carbon nanotubes have the advantage to cross the plasma membrane without damaging the cells, improving the biological effect of a drug and reducing its side effects. In the present study, the effect of metformin and metformin-conjugated nanotubes was investigated on blood glucose level in the streptozotocin-induced male diabetic rats. Diabetes in the animals was induced with a single dose of streptozotocin (60 mg/kg; i.p.) and after 3 days the blood glucose was analyzed. Animals showing fasting blood glucose higher than 250 mg/dL were considered as diabetic rats. The animals were treated with metformin and metformin-conjugated nanotubes (150 mg/kg; p.o.) daily and every 48-h for 1 week. Changes in animals' serum blood glucose level were evaluated daily during the treatment period. The results of this study showed that metformin reduced blood glucose levels in diabetic animals. Metformin-conjugated nanotubes significantly reduced the blood glucose levels in diabetic rats (p < 0.01). There was no significant difference in blood glucose level between metformin and metformin-conjugated nanotubes groups (p > 0.05). However, when both formulations of metformin were administered every 48-h, metformin-conjugated nanotubes reduced glycaemia for a longer time than metformin alone (p < 0.001). This study showed that the metformin-conjugated nanotubes would be able to reduce the blood glucose, prolong drug delivery and efficacy duration in animals which were treated with metformin-conjugated nanotubes compared with metformin alone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Schwefelsäure, 99.999%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydrid, 60 % dispersion in mineral oil
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydrid, dry, 90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Thionylchlorid, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Thionylchlorid, reagent grade, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumsulfat, ≥99.0%, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N
Preise und Verfügbarkeit sind derzeit nicht verfügbar.