Merck
  • Comparative transcriptomics reveals key differences in the response to milk oligosaccharides of infant gut-associated bifidobacteria.
Comparative transcriptomics reveals key differences in the response to milk oligosaccharides of infant gut-associated bifidobacteria.
Scientific reports (2015-09-05)
Daniel Garrido, Santiago Ruiz-Moyano, Danielle G Lemay, David A Sela, J Bruce German, David A Mills
ZUSAMMENFASSUNG
Breast milk enhances the predominance of Bifidobacterium species in the infant gut, probably due to its large concentration of human milk oligosaccharides (HMO). Here we screened infant-gut isolates of Bifidobacterium longum subsp. infantis and Bifidobacterium bifidum using individual HMO, and compared the global transcriptomes of representative isolates on major HMO by RNA-seq. While B. infantis displayed homogeneous HMO-utilization patterns, B. bifidum were more diverse and some strains did not use fucosyllactose (FL) or sialyllactose (SL). Transcriptomes of B. bifidum SC555 and B. infantis ATCC 15697 showed that utilization of pooled HMO is similar to neutral HMO, while transcriptomes for growth on FL were more similar to lactose than HMO in B. bifidum. Genes linked to HMO-utilization were upregulated by neutral HMO and SL, but not by FL in both species. In contrast, FL induced the expression of alternative gene clusters in B. infantis. Results also suggest that B. bifidum SC555 does not utilize fucose or sialic acid from HMO. Surprisingly, expression of orthologous genes differed between both bifidobacteria even when grown on identical substrates. This study highlights two major strategies found in Bifidobacterium species to process HMO, and presents detailed information on the close relationship between HMO and infant-gut bifidobacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Kohlenstoffdioxid, ≥99.8%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Stickstoff, ≥99.998%
SAFC
L-Cystein
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Cystein, ≥97%, FG
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.9 atom % 12C
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.99 atom % 12C
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O