Merck
  • Self-assembly micelles with lipid core of cholesterol for docetaxel delivery to B16F10 melanoma and HepG2 cells.
Self-assembly micelles with lipid core of cholesterol for docetaxel delivery to B16F10 melanoma and HepG2 cells.
Journal of liposome research (2014-10-03)
Jaleh Varshosaz, Somayeh Taymouri, Farshid Hassanzadeh, Shaghaiegh Haghjooy Javanmard, Mahboobeh Rostami
ZUSAMMENFASSUNG
The objective of the present study was to prepare a micellar polymeric carrier for the delivery of Docetaxel (DTX) as a kind of polysorbate free preparation. Pluronic F127 (PF127) was conjugated to cholesterol (Chol) via succinyl linkage and characterized by FTIR and HNMR. DTX-loaded polymeric micelles were prepared via film hydration method. Physicochemical properties of micelles including particle size, zeta potential, drug loading and release efficiency were studied. The critical micelle concentration (CMC) was determined using pyrene as a hydrophobic fluorescent probe. In vitro cytotoxicity of micelles was evaluated in B16F10 melanoma cells and HepG2 cell line. The FTIR and HNMR spectroscopy methods confirmed the conjugation of PF127 to cholesterol via succinyl linkage. The micelles were spherical under scanning electron microscope (SEM) with the mean particle size of 248.2 ± 8-278.8 ± 12.3 nm and zeta potential ranging from -17.2 ± 8.7 to -28.4 ± 12.7 mV. Drug loading efficiency was higher than 98%. The in vitro release study showed the sustained release behavior of DTX within 144 h. The CMC of the micelles was about 41.67 ± 0.17 µg/ml, which was significantly lower than the CMC of pure PF127 micelles. Compared with the free drug, DTX-loaded micelles showed higher cytotoxicity against B16F10 melanoma and HepG2 cell lines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Bernsteinsäureanhydrid, ≥99% (GC)
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Pyren, 98%
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Pyren, sublimed grade, 99%
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.