Merck
  • Sustained release of TGF-β1 from biodegradable microparticles prepared by a new green process in CO2 medium.
Sustained release of TGF-β1 from biodegradable microparticles prepared by a new green process in CO2 medium.
International journal of pharmaceutics (2015-07-26)
Amin Swed, Thomas Cordonnier, Alison Dénarnaud, Cécile Boyer, Jérôme Guicheux, Pierre Weiss, Frank Boury
ZUSAMMENFASSUNG
The aim of this work was to encapsulate transforming growth factor β1 (TGF-β1) into PLGA microparticles for regenerative medicine applications. TGF-β1 was firstly precipitated to ensure its stability during subsequent encapsulation within microparticles. A novel emulsification/extraction process in CO2 medium under mild conditions of pressure and temperature was used to encapsulate the protein. Interestingly, non-volatile injectable solvents, isosorbide dimethyl ether (DMI) and glycofurol (GF), were employed to precipitate the protein and to dissolve the polymer. Good encapsulation efficiency was obtained with preserved bioactivity of the protein. The microparticles were characterized in terms of size and zeta potential. In addition, the morphology and surface properties were determined using scanning electron microscopy (SEM) and atomic force microscopy (AFM) respectively. In vitro release study of the protein from microparticles was presented to assess the capacity of these systems to control the protein release. Moreover, cytotoxicity study was performed and showed an excellent cytocompatibility of the obtained microparticles. Thus, we described an effective and original process for TGF-β1 encapsulation into PLGA microparticles. The obtained polymeric carriers could be used in many biomedical applications and were more specifically developed for cartilage regeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tetraethylenglycol, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Glycin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isosorbiddimethylether, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.