Merck
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Autophagy (2015-06-30)
Chi G Weindel, Lauren J Richey, Silvia Bolland, Abhiruchi J Mehta, John F Kearney, Brigitte T Huber
ZUSAMMENFASSUNG
Systemic lupus erythematosus (SLE) is a heterogeneous autoimmune disease, defined by loss of B cell self-tolerance that results in production of antinuclear antibodies (ANA) and chronic inflammation. While the initiating events in lupus development are not well defined, overexpression of the RNA-recognizing toll-like receptor (TLR)7 has been linked to SLE in humans and mice. We postulated that autophagy plays an essential role in TLR7 activation of B cells for the induction of SLE by delivering RNA ligands to the endosomes, where this innate immune receptor resides. To test this hypothesis, we compared SLE development in Tlr7 transgenic (Tg) mice with or without B cell-specific ablation of autophagy (Cd19-Cre Atg5(f/f)). We observed that in the absence of B cell autophagy the 2 hallmarks of SLE, ANA and inflammation, were eliminated, thus curing these mice of lupus. This was also evident in the significantly extended survival of the autophagy-deficient mice compared to Tlr7.1 Tg mice. Furthermore, glomerulonephritis was ameliorated, and the serum levels of inflammatory cytokines in the knockout (KO) mice were indistinguishable from those of control mice. These data provide direct evidence that B cells require TLR7-dependent priming through an autophagy-dependent mechanism before autoimmunity is induced, thereafter involving many cell types. Surprisingly, hyper-IgM production persisted in Tlr7.1 Tg mice in the absence of autophagy, likely involving a different activation pathway than the production of autoantibodies. Furthermore, these mice still presented with anemia, but responded with a striking increase in extramedullary hematopoiesis (EMH), possibly due to the absence of pro-inflammatory cytokines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
L-Glutamin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Formalinlösung, neutral gepuffert, 10 %, histological tissue fixative
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.