Merck
  • Capillary coated with graphene oxide as stationary phase for the separation of brucine and strychnine by capillary electrophoresis.
Capillary coated with graphene oxide as stationary phase for the separation of brucine and strychnine by capillary electrophoresis.
Journal of chromatographic science (2014-07-06)
Jian Li, Nengsheng Ye, Chong Gao, Tingting Zhou, Jichao Ma
ZUSAMMENFASSUNG
A new capillary electrophoresis (CE) method was developed by using graphene oxide (GO) as a stationary phase for the separation of brucine and strychnine. The separation performance, reproducibility and stability of GO-coated capillary were investigated for the analysis of brucine and strychnine. After optimization of the separation conditions, a phosphate solution (40 mM, pH 7.0) containing 25% (v/v) acetonitrile was selected as the running buffer. Compared with uncoated capillary, higher separation efficiency was achieved by GO-coated capillary as a result of the increasing interactions between the analytes and the stationary phase of capillary. The linear ranges of these two alkaloids were 4.0-100.0 μg mL(-1) with a satisfied correlation coefficients (R > 0.9994), and this novel method provided an efficient separation of brucine and strychnine as well as a good reproducibility and stability. Finally, the developed method was successfully applied for the determination of these two alkaloids in a pharmaceutical formulation of traditional Chinese medicines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natriumphosphat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumphosphat, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Natriumphosphat, zweibasisch -Lösung, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets