Merck
  • On-line liquid chromatography-gas chromatography: A novel approach for the analysis of phytosterol oxidation products in enriched foods.
On-line liquid chromatography-gas chromatography: A novel approach for the analysis of phytosterol oxidation products in enriched foods.
Journal of chromatography. A (2015-04-22)
Birgit Scholz, Stefan Wocheslander, Vera Lander, Karl-Heinz Engel
ZUSAMMENFASSUNG
A novel methodology for the automated qualitative and quantitative determination of phytosterol oxidation products in enriched foods via on-line liquid chromatography-gas chromatography (LC-GC) was established. The approach is based on the LC pre-separation of acetylated phytosterols and their corresponding oxides using silica as stationary phase and a mixture of n-hexane/methyl tert-butyl ether/isopropanol as eluent. Two LC-fractions containing (i) 5,6-epoxy- and 7-hydroxyphytosterols, and (ii) 7-ketophytosterols are transferred on-line to the GC for the analysis of their individual compositions on a medium polar trifluoropropylmethyl polysiloxane capillary column. Thus, conventionally employed laborious off-line purification and enrichment steps can be avoided. Validation data, including recovery, repeatability, and reproducibility of the method, were elaborated using an enriched margarine as example. The margarine was subjected to a heating procedure in order to exemplarily monitor the formation of phytosterol oxidation products. Quantification was performed using on-line LC-GC-FID, identification of the analytes was based on on-line LC-GC-MS. The developed approach offers a new possibility for the reliable and fast analysis of phytosterol oxidation products in enriched foods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Essigsäureanhydrid, ACS reagent, ≥98.0%
Sigma-Aldrich
Natriummethoxid -Lösung, 25 wt. % in methanol
Sigma-Aldrich
Natriummethoxid, reagent grade, 95%, powder
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Essigsäureanhydrid, 99.5%
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
Natriummethoxid -Lösung, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Campesterol, ~65%
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
2-Hydroxybuttersäure, 97%