Merck
  • 3D imaging of cell interactions with electrospun PLGA nanofiber membranes for bone regeneration.
3D imaging of cell interactions with electrospun PLGA nanofiber membranes for bone regeneration.
Acta biomaterialia (2015-09-09)
Urszula Stachewicz, Tuya Qiao, Simon C F Rawlinson, Filipe Veiga Almeida, Wei-Qi Li, Michael Cattell, Asa H Barber
ZUSAMMENFASSUNG
The interaction between resident cells and electrospun nanofibers is critical in determining resultant osteoblast proliferation and activity in orthopedic tissue scaffolds. The use of techniques to evaluate cell-nanofiber interactions is critical in understanding scaffold function, with visualization promising unparalleled access to spatial information on such interactions. 3D tomography exploiting focused ion beam (FIB)-scanning electron microscopy (SEM) was used to examine electrospun nanofiber scaffolds to understand the features responsible for (osteoblast-like MC3T3-E1 and UMR106) cell behavior and resultant scaffold function. 3D imaging of cell-nanofiber interactions within a range of electrospun poly(d,l-lactide-co-glycolide acid) (PLGA) nanofiber scaffold architectures indicated a coherent interface between osteoblasts and nanofiber surfaces, promoting osteoblast filopodia formation for successful cell growth. Coherent cell-nanofiber interfaces were demonstrated throughout a randomly organized and aligned nanofiber network. Gene expression of UMR106 cells grown on PLGA fibers did not deviate significantly from those grown on plastic, suggesting maintenance of phenotype. However, considerably lower expression of Ibsp and Alpl on PLGA fibers might indicate that these cells are still in the proliferative phase compared with a more differentiated cell on plastic. This work demonstrates the synergy between designing electrospun tissue scaffolds and providing comprehensive evaluation through high resolution imaging of resultant 3-dimensional cell growth within the scaffold. Membranes made from electrospun nanofibers are potentially excellent for promoting bone growth for next-generation tissue scaffolds. The effectiveness of an electrospun membrane is shown here using high resolution 3D imaging to visualize the interaction between cells and the nanofibers within the membrane. Nanofibers that are aligned in one direction control cell growth at the surface of the membrane whereas random nanofibers cause cell growth into the membrane. Such observations are important and indicate that lateral cell growth at the membrane surface using aligned nanofibers could be used for rapid tissue repair whereas slower but more extensive tissue production is promoted by membranes containing random nanofibers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
SAFC
L-Glutamin
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamid, synthesis grade, ≥95%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Sigma-Aldrich
Osmiumtetroxid -Lösung, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxan-2,5-dion, 99%
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed