Merck
  • Self assembled nano fibers of betulinic acid: A selective inducer for ROS/TNF-alpha pathway mediated leukemic cell death.
Self assembled nano fibers of betulinic acid: A selective inducer for ROS/TNF-alpha pathway mediated leukemic cell death.
Bioorganic chemistry (2015-10-16)
Sandeep Kumar Dash, Sourav Chattopadhyay, Shib Shankar Dash, Satyajit Tripathy, Balaram Das, Santanu Kar Mahapatra, Braja Gopal Bag, Parimal Karmakar, Somenath Roy
ZUSAMMENFASSUNG
The main complication in betulinic acid (BA) based drug delivery has been observed due to its bulk structure. The present study demonstrates the potential effects of self assembled nano size betulinic acid (SA-BA) by treating human leukemic cell lines (KG-1A and K562) and its normal counterpart. Self assembly property of BA was investigated using SEM and DLS study which showed that the BA forms fibrous structure having few nanometers in diameter. Selective anti-leukemic efficacy study of SA-BA was investigated by cell viability assay. Mode of leukemic cell death and probable pathway of apoptosis was monitored by measuring ROS level, pro and anti-inflammatory cytokine status and expression of caspase-8 and caspage-3 by immunocytochemistry. Higher efficacy of SA-BA over non-assemble BA was monitored toward cancer cells, with no relevant toxicity to normal blood cells. SA-BA facilitated reactive oxygen species (ROS) mediated leukemic cell death, confirmed by pre-treatment of N-acetyl-L-cysteine. Induction of apoptosis by SA-BA treatment increased pro-inflammatory cytokines, specifically potentiated TNF-α mediated cell death, confirmed by expression of caspase-8 and caspage-3 by immunocytochemistry. This study explored the better anti-leukemic efficacy of SA-BA over BA and this modification will enrich the use of BA in cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
N-Acetyl-L-Cystein, suitable for cell culture, BioReagent
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%