Selecione um tamanho
| Tamanho/SKU | Disponibilidade | Preço |
|---|---|---|
1 ea | Verificar disponibilidade no carrinho | R$ 1.654,00 |
Sobre este item
R$ 1.654,00
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P140010M
capacity
10 mL , 10 mL
fitting
GL 14
General description
Legal Information
1 of 1
Este Item | |||
|---|---|---|---|
| description 10 mL, joint: ST/NS 24/40, porosity: medium, GL 14 | description 100 mL, joint: ST/NS 24/40, porosity: medium, GL 25 | description 50 mL, joint: ST/NS 24/40, porosity: medium, GL 25 | description 25 mL, joint: ST/NS 24/40, porosity: medium, GL 25 |
| manufacturer/tradename Synthware P140010M | manufacturer/tradename Synthware P140100M | manufacturer/tradename Synthware P140050M | manufacturer/tradename Synthware P140025M |
| capacity 10 mL, 10 mL | capacity 100 mL , 100 mL | capacity 50 mL, 50 mL | capacity 25 mL , 25 mL |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| fitting GL 14 | fitting GL 25 | fitting GL 25 | fitting GL 25 |
Still not finding the right product?
Explore all of our products under Synthware™ peptide synthesis vessel with fritted disc, T-bore PTFE stopcock and vacuum take-off
Escolha uma das versões mais recentes:
Já possui este produto?