MXRPCUK
Propriedades
250 V, 10 A
SP1
for use with MOBIUS® Power MIX - all sizes
Descrição
Escolha uma das versões mais recentes:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.
Categorias relacionadas
Bioprocessing Systems
Buffer & Media Prep
Pharma & Biopharma Manufacturing
Single-Use Mixers