Merck
Pular para
GE11-0012-54
Tricorn 10 Coarse Filter Kit
Faça login para ver os preços organizacionais e de contrato.
Sobre este item
UNSPSC Code:
41106500
description
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography
feature
Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM) , Wetted Materials: Ethylene propylene diene (EPDM), Polyethylene (PE)
manufacturer/tradename
Cytiva 11-0012-54
parameter
max. temp. (60 °C), 4 °C min. temp.
Escolha uma das versões mais recentes:
Certificados de análise (COA)