Merck
  • Pharmacokinetics of repeated sodium salicylate administration to laying hens: evidence for time dependent increase in drug elimination from plasma and eggs.
Pharmacokinetics of repeated sodium salicylate administration to laying hens: evidence for time dependent increase in drug elimination from plasma and eggs.
PloS one (2015-04-22)
Błażej Poźniak, Tomasz Grabowski, Karolina Motykiewicz-Pers, Kamila Bobrek, Lech Rak, Katarzyna Bobusia, Andrzej Gaweł, Marcin Świtała
ZUSAMMENFASSUNG
Salicylates were the first non-steroid anti-inflammatory drugs (NSAIDs) to be used in any species and are still widely used in humans and livestock. However, the data on their pharmacokinetics in animals is limited, especially after repeated administration. Evidence exist that in chickens (Gallus gallus) salicylate (SA) may induce its own elimination. The aim of this study was to investigate salicylate pharmacokinetics and egg residues during repeated administration of sodium salicylate (SS) to laying hens. Pharmacokinetics of SA was assessed during 14 d oral administration of SS at daily doses of 50 mg/kg and 200 mg/kg body weight to laying hens. On the 1st, 7th and 14th d a 24 h-long pharmacokinetic study was carried out, whereas eggs were collected daily. Salicylate concentrations in plasma and eggs were determined using high-performance liquid chromatography with ultraviolet detection and pharmacokinetic variables were calculated using a non-compartmental model. Mean residence time (MRT), minimal plasma concentration (Cmin, C16h) and elimination half-life (T1/2el) of SA showed gradual decrease in layers administered with a lower dose. Total body clearance (ClB) increased. Layers administered with the higher dose showed a decrease only in the T1/2el. In the low dose group, SA was found only in the egg white and was low throughout the experiment. Egg whites from the higher dose group showed initially high SA levels which significantly decreased during the experiment. Yolk SA levels were lower and showed longer periods of accumulation and elimination. Repeated administration of SS induces SA elimination, although this effect may differ depending on the dose and production type of a chicken. Decreased plasma drug concentration may have clinical implications during prolonged SS treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, ≥99.9% (GC)
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Acetonitril, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitril, ≥99.5% (GC)
Sigma-Aldrich
Trichloressigsäure, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, ≥99.8% (GC)