Merck
  • Protective efficacy of ursodeoxycholic acid nanoparticles in animal model of inflammatory bowel disease.
Protective efficacy of ursodeoxycholic acid nanoparticles in animal model of inflammatory bowel disease.
Journal of microencapsulation (2014-06-26)
Amit Kumar De, Santanu Sana, Sriparna Datta, Arup Mukherjee
ZUSAMMENFASSUNG
Abstract Context: Ursodeoxycholic acid (UDCA) exerts dose-dependent chemoprevention in colonic inflammation. Polycationic UDCA nanoparticles (UNPs) are envisaged for solubility enhancement and site directed drug delivery. The objective was to study the ameliorative efficacy of UNPs through localized delivery of the drug. UNPs were prepared through nanoprecipitation technique. Particle size, morphology, in vitro drug release and in vivo protective efficacy in inflammatory bowel disease (IBD) of these nanoparticles were studied. The average particle size was around 100 nm, and the average drug encapsulation was about 99%. In vitro drug release study shows optimal drug release in simulated colonic fluid. The lowering of tissue nitric oxide, malondialdehyde, myeloperoxidase and histology of the colon tissue supported the protective efficacy of the nanoparticles. This study presents the improved efficacy of UNPs in animal model of IBD due to complete release of drug at the desired site of action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Wasserstoffperoxid-Lösung, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, purum p.a., ≥35% (RT)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
o-Dianisidin -dihydrochlorid, ≥95%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥96.0% (AT)
Supelco
Sulfanilamid Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard