Merck
Placental syndecan-1 and sulphated glycosaminoglycans are decreased in preeclampsia.
Journal of perinatal medicine (2013-11-14)
Narinda Heyer-Chauhan, Ihinosen J Ovbude, Anne A Hills, Mark H Sullivan, Frank A Hills
ZUSAMMENFASSUNG
Glycosaminoglycans are found in extracellular matrix and on the cell surface in the form of proteoglycans. There is evidence that these molecules regulate biological processes, including cell survival, migration and angiogenesis. Preeclampsia is a common pregnancy disorder associated with insufficient placental development. This study aimed to determine the concentrations of glycosaminoglycans and the proteoglycan syndecan-1 within villous trophoblast and to investigate changes associated with preeclampsia. Seventy-five placental samples collected from third trimester singleton pregnancies were divided into term placentas following labour onset, gestational age-matched placentas prior to labour onset and preterm placentas. Preterm placentas were divided into three gestational age-matched groups, spontaneous preterm labour, preterm premature rupture of membranes (PPROM) and preterm preeclampsia. Sulphated glycosaminoglycan (sGAG) concentrations in placental extracts were quantified using a modified 1,9-dimethylmethylene blue assay. Syndecan-1 expression was localised using immunohistochemistry and concentrations in placental extracts determined by immunoassay. Preterm placentas had significantly lower sGAG concentrations compared to term tissues and concentrations were significantly lower in preeclampsia compared to spontaneous preterm labour (medians 5.80 and 10.0 μg/mg protein respectively, P<0.05). Syndecan-1 expression was localised to syncytiotrophoblast and median concentrations were lower in preeclampsia compared to PPROM material (preeclampsia median = 41.7, PPROM 74.4 ng/mg tissue) but not significantly different to concentrations in spontaneous preterm labour. Multivariate analysis revealed that decreased sGAG and syndecan-1 in preeclampsia were independent of labour, gestational age and birthweight centile. These findings may provide insights into a role for these molecules in the pathophysiology of preeclampsia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ameisensäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
o-Xylen, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Ameisensäure, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Guanidin -hydrochlorid, ≥99% (titration), organic base and chaeotropic agent
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Guanidin -hydrochlorid, ≥98%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%