Merck
  • Hepatoprotective and antioxidant activity of Melaleuca styphelioides on carbon tetrachloride-induced hepatotoxicity in mice.
Hepatoprotective and antioxidant activity of Melaleuca styphelioides on carbon tetrachloride-induced hepatotoxicity in mice.
Pharmaceutical biology (2014-09-23)
Eman Al-Sayed, Naglaa M El-Lakkany, Sayed H Seif El-Din, Abdel-Nasser A Sabra, Olfat A Hammam
ZUSAMMENFASSUNG
Liver disease is a serious problem. Polyphenolic compounds have marked antioxidant effect and can prevent the liver damage caused by free radicals. In vitro studies have revealed the strong antioxidant activity of an ellagitannin-rich plant, namely, Melaleuca styphelioides Sm. (Myrtaceae). In view of the limited therapeutic options available for the treatment of liver diseases, the hepatoprotective potential of the methanol extract of M. styphelioides leaves (MSE) was investigated against CCl4-induced liver injury in mice. MSE was administered (500 and 1000 mg/kg/d p.o.) along with CCl4 for 6 weeks. Alanine aminotransferase (ALT), aspartate aminotransferase (AST), and alkaline phosphatase (ALP) were determined in the serum. Glutathione (GSH), superoxide dismutase (SOD), glutathione peroxidase (GPx), glutathione reductase (GR), glutathione transferase (GST), and malondialdehyde (MDA) were estimated in the liver homogenate. The bioactive components of MSE were identified by NMR, UV and HRESI-MS/MS data. MSE treatment (500 and 1000 mg/kg/d) markedly inhibited the CCl4-induced increase in the levels of AST (31 and 38%), ALT (29 and 32%), ALP (13 and 19%), and MDA (22 and 37%) at the tested doses, respectively. MSE treatment markedly increased the levels of GSH (29 and 57%) and antioxidant enzymes compared with the CCl4-treated group. MSE was more effective than silymarin in restoring the liver architecture and reducing the fatty changes, central vein congestion, Kupffer cell hyperplasia, inflammatory infiltration, and necrosis induced by CCl4. The LD50 of MSE was more than 5000 mg/kg. MSE confers potent antioxidant and hepatoprotective effects against CCl4-induced toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Aluminiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Aluminiumchlorid, reagent grade, 98%
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
(+)-Natrium-L-ascorbat, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
Aluminiumchlorid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Aluminiumchlorid, 99.99% trace metals basis
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminiumchlorid, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminiumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Natriumascorbat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminiumchlorid -Lösung, 1.0 M in nitrobenzene
Sigma-Aldrich
Trichloressigsäure, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Natriumascorbat, European Pharmacopoeia (EP) Reference Standard