Merck
  • Steroidal constituents from the edible sea urchin Diadema savignyi Michelin induce apoptosis in human cancer cells.
Steroidal constituents from the edible sea urchin Diadema savignyi Michelin induce apoptosis in human cancer cells.
Journal of medicinal food (2014-09-12)
Nguyen Phuong Thao, Bui Thi Thuy Luyen, Eun Ji Kim, Jung Il Kang, Hee Kyoung Kang, Nguyen Xuan Cuong, Nguyen Hoai Nam, Phan Van Kiem, Chau Van Minh, Young Ho Kim
ABSTRACT
Bioassay-directed fractionation and purification were used to isolate 12 steroids (1-12) from a CH(2)Cl(2) extract of the edible Vietnamese sea urchin Diadema savignyi Michelin. The cytotoxic activity of the CH(2)Cl(2) extract and 12 steroids was evaluated in three human cancer cell lines (HL-60, PC-3, and SNU-C5). Relative to the effects of the positive control, mitoxantrone, the CH(2)Cl(2) extract (with an inhibitory concentration of 50% [IC(50)] values ranging from 1.37±0.15 to 3.11±0.15 μg/mL) and compounds 2 (with IC(50) values ranging from 5.29±0.11 to 6.80±0.67 μM) and 11 (with IC(50) values ranging from 4.95±0.07 to 6.99±0.28 μM) exhibited potent cytotoxic effects against all three tested human cancer cell lines. In addition, the CH(2)Cl(2) extract and compounds 2 and 11 were found to induce apoptosis. The induction of apoptosis was accompanied by alterations of the apoptosis-related protein expression, inactivation of ERK1/2 mitogen-activated protein kinase signaling, and decreased c-Myc expression. These data suggest that compounds 2 and 11 from the edible sea urchin D. savignyi may have potential for the treatment of colon cancer, leukemia, and prostate cancer as complementary cancer remedies.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%