Merck
  • Analysis of epigenetic modifications of chromatin at specific gene loci by native chromatin immunoprecipitation of nucleosomes isolated using hydroxyapatite chromatography.
Analysis of epigenetic modifications of chromatin at specific gene loci by native chromatin immunoprecipitation of nucleosomes isolated using hydroxyapatite chromatography.
Nature protocols (2008-03-08)
Marjorie Brand, Shravanti Rampalli, Chandra-Prakash Chaturvedi, F Jeffrey Dilworth
ABSTRACT
Chromatin immunoprecipitation (ChIP) is routinely used to examine epigenetic modification of histones at specific genomic locations. However, covalent modifications of histone tails can serve as docking sites for chromatin regulatory factors. As such, association of these regulatory factors with chromatin could cause steric hindrance for antibody recognition, resulting in an underestimation of the relative enrichment of a given histone modification at specific loci. To overcome this problem, we have developed a native ChIP protocol to study covalent modification of histones that takes advantage of hydroxyapatite (HAP) chromatography to wash away chromatin-associated proteins before the immunoprecipitation of nucleosomes. This fast and simple procedure consists of five steps: nuclei isolation from cultured cells; fragmentation of chromatin using MNase; purification of nucleosomes using HAP; immunoprecipitation of modified nucleosomes; and qPCR analysis of DNA associated with modified histones. Nucleosomes prepared in this manner are free of contaminating proteins and permit an accurate evaluation of relative abundance of different covalent histone modifications at specific genomic loci. Completion of this protocol requires approximately 1.5 d.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glicerolo, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acido etilendiamminotetraacetico, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Saccarosio, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Fenil metansolfonile fluoruro, ≥98.5% (GC)
Sigma-Aldrich
Bromuro di etidio, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Cloruro di calcio, Molecular Biology, ≥99.0%
Sigma-Aldrich
Acido desossi-ribonucleico
Sigma-Aldrich
Proteinasi K, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Cloruro di magnesio, BioXtra, ≥99.0%
Sigma-Aldrich
Fosfato di sodio, BioXtra, ≥99.0%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Saccarosio, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Nuclease micrococcal from Staphylococcus aureus, 100-300 units/mg protein