Merck
  • Optimal design and experimental validation of a simulated moving bed chromatography for continuous recovery of formic acid in a model mixture of three organic acids from Actinobacillus bacteria fermentation.
Optimal design and experimental validation of a simulated moving bed chromatography for continuous recovery of formic acid in a model mixture of three organic acids from Actinobacillus bacteria fermentation.
Journal of chromatography. A (2014-09-23)
Chanhun Park, Hee-Geun Nam, Ki Bong Lee, Sungyong Mun
ABSTRACT
The economically-efficient separation of formic acid from acetic acid and succinic acid has been a key issue in the production of formic acid with the Actinobacillus bacteria fermentation. To address this issue, an optimal three-zone simulated moving bed (SMB) chromatography for continuous separation of formic acid from acetic acid and succinic acid was developed in this study. As a first step for this task, the adsorption isotherm and mass-transfer parameters of each organic acid on the qualified adsorbent (Amberchrom-CG300C) were determined through a series of multiple frontal experiments. The determined parameters were then used in optimizing the SMB process for the considered separation. During such optimization, the additional investigation for selecting a proper SMB port configuration, which could be more advantageous for attaining better process performances, was carried out between two possible configurations. It was found that if the properly selected port configuration was adopted in the SMB of interest, the throughout and the formic-acid product concentration could be increased by 82% and 181% respectively. Finally, the optimized SMB process based on the properly selected port configuration was tested experimentally using a self-assembled SMB unit with three zones. The SMB experimental results and the relevant computer simulation verified that the developed process in this study was successful in continuous recovery of formic acid from a ternary organic-acid mixture of interest with high throughput, high purity, high yield, and high product concentration.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Solfato di sodio, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
Solfato di sodio, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Supelco
Formato di ammonio, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formato di ammonio, reagent grade, 97%
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acido metansolfonico, ≥99.0%
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Acido acetico, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acido formico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formato di ammonio, ≥99.995% trace metals basis
Sigma-Aldrich
Acido formico, ACS reagent, ≥88%
Sigma-Aldrich
Solfato di sodio, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Solfato di sodio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Acido acetico, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formato di ammonio, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acido succinico, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
USP
Acido acetico, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acido succinico, ACS reagent, ≥99.0%
Sigma-Aldrich
Solfato di sodio, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Supelco
Acido acetico, analytical standard
Sigma-Aldrich
Solfato di sodio, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Acido formico, ≥95%, FCC, FG