Merck
  • Effects of 4-nonylphenol isomers on cell receptors and mitogen-activated protein kinase pathway in mouse Sertoli TM4 cells.
Effects of 4-nonylphenol isomers on cell receptors and mitogen-activated protein kinase pathway in mouse Sertoli TM4 cells.
Toxicology (2014-09-23)
Xiaozhen Liu, Shaoping Nie, Yangjie Chen, Danfei Huang, Mingyong Xie
ABSTRACT
In the present study, experiments were performed to investigate the effects of nonylphenol (NP) isomers (4-[1,2, 4-trimethylhexyl]-phenol (NP41), 4-[1,2, 5-trimethylhexyl]-phenol (NP42)) on Sertoli TM4 cells. NP41 decreased mRNA expression levels of androgen receptor and toll-like receptor (TLR)-4 in 20-40μM (P<0.05), and increased mRNA levels of estrogen receptor (ER)-α and progesterone receptor in 1-40μM (P<0.05). NP42 treatment only evoked significant decrease in mRNA expression levels of ER-α in 20-40μM (P<0.05). Similarly, NP41 (1-40μM) drastically increased the protein expression of ER-α, which was significantly decreased in 20-40μM NP42 groups (P<0.01). Both NP41 and NP42 showed no effect on the expression of ER-β. Protein levels of follicle stimulating hormone receptor were increased significantly in high concentrations of NP41 (40μM) and NP42 (10-40μM) challenged cells. Furthermore, NP41 and NP42 showed various effects on the expression of junction-associated molecules and inhibin B secretion in TM4 cells. Additionally, activation of JNK1/2 pathway was induced by NP41 and NP42. However, ERK1/2 and p38 pathways were inhibited in TM4 cells exposed to low concentrations of NP41 (0.1-20μM) and NP42 (0.1-1μM), and high concentrations of NP41 (40μM) and NP42 (10-40μM) resulted in a return of p-ERK1/2 and p-p38 to control levels. We proposed that molecular mechanism of reproductive damage in Sertoli cells induced by NPs may be mediated by cell receptors and/or cell signaling pathways, and the effects may be related to the structure of NP isomer.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Streptomicina, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomicina, powder
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Streptomicina, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Acido bicinconinico, ≥98% (HPLC)
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Streptomicina, European Pharmacopoeia (EP) Reference Standard