Merck
Hepatitis B virus e antigen (HBeAg) may have a negative effect on dendritic cell generation.
Immunobiology (2014-08-26)
Ibrahim Hatipoglu, Duygu Ercan, Ceyda Acilan, Aynur Basalp, Deniz Durali, Ahmet Tarik Baykal
ABSTRACT
Hepatitis B virus (HBV) continues to be a serious worldwide health problem despite the use of protective HBV vaccines and therapeutic regimens against chronic HBV infection. Chronic HBV patients cannot induce sufficient immune responses against the virus. HBV and its antigens are believed to suppress immune responses during chronic infection. Hence, studying the role of HBV in immune suppression is very important for the development of alternative therapeutic strategies for HBV infections. In the present study, we investigated the effect of Hepatitis B virus e antigen (HBeAg) on the generation of bone marrow derived dendritic cells (BMDCs) and the stimulation of plasmacytoid DCs (pDCs). In the presence of HBeAg, the ratio of BMDCs was decreased, but the ratio of CD11b(+)Ly6G(+) immature myeloid cells was increased. The expression of 47 proteins was also changed during HBeAg treatment; however, CpG-induced MHC-II expression on pDCs was not affected. Our results indicate that HBeAg may have a negative effect on the generation of DCs from bone morrow precursors.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Bicarbonato di sodio, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Bicarbonato di sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Bicarbonato di sodio, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acido formico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Acido formico, ACS reagent, ≥88%
Sigma-Aldrich
3-Indoleacetic acid, 98%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Bicarbonato di sodio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Bicarbonato di sodio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Soluzione tampone HEPES, 1 M in H2O
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)