Merck
  • Simultaneous determination of osthole, bergapten and isopimpinellin in rat plasma and tissues by liquid chromatography-tandem mass spectrometry.
Simultaneous determination of osthole, bergapten and isopimpinellin in rat plasma and tissues by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-23)
Jing Li, Bo Ma, Qi Zhang, Xiaojing Yang, Jingjing Sun, Bowen Tang, Guangbo Cui, Di Yao, Lei Liu, Guiying Gu, Jianwei Zhu, Ping Wei, Pingkai Ouyang
ABSTRACT
A highly selective and sensitive method for simultaneous quantitation of osthole, bergapten and isopimpinellin in rat plasma and tissues was developed by liquid chromatography-tandem quadrupole mass spectrometry (LC-MS/MS). After liquid-liquid extraction of samples with methyl tert-butyl ether, the analytes and dextrorphan (internal standard, IS) were separated by a Hypersil GOLD AQ C18 column with gradient elution of acetonitrile and water containing 0.5‰ formic acid. Three determinands were detected using an electrospray ionization (ESI) tandem mass spectrometry in the multiple reaction monitoring (MRM) modes with positive electrospray ionization. Calibration curves were recovered over the concentration ranges of 1-200 ng/ml, 1-500 ng/ml, 0.25-200 ng/ml for osthole, bergapten and isopimpinellin in plasma; 1-100 ng/ml, 1-500 ng/ml, 0.5-100 ng/ml for osthole, bergapten and isopimpinellin in tissues, respectively. The intra-day precision (R.S.D.) was within 13.90% and the intra-day accuracy (R.E.) was within -6.27 to 6.84% in all biological matrixes. The inter-day precision (R.S.D.) was less than 13.66% and the inter-day accuracy (R.E.) was within -10.64 to 13.04%. Then the method was successfully applied to investigate plasma pharmacokinetic study and tissue distribution of osthole, bergapten and isopimpinellin in rats after oral administration of Fructus Cnidii extraction, especially for testis/uterus tissue distribution. The results demonstrated that osthole, bergapten and isopimpinellin were absorbed and eliminated rapidly with wide distributions in rats. Distribution data of these three bioactive components in testis/uterus tissues could offer useful information for the further preclinical and clinical studies of Fructus Cnidii in the treatment of genital system disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
ter-butil metil etere, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
ter-butil metil etere, ACS reagent, ≥99.0%
Sigma-Aldrich
ter-butil metil etere, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acido formico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acido formico, ACS reagent, ≥88%
Sigma-Aldrich
ter-butil metil etere, reagent grade, ≥98%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
ter-butil metil etere, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acido formico, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
ter-butil metil etere, analytical standard
Supelco
ter-butil metil etere, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
ter-butil metil etere, reagent grade, 98%
Sigma-Aldrich
Dextrorphan