Merck
  • Comparison of peak-picking workflows for untargeted liquid chromatography/high-resolution mass spectrometry metabolomics data analysis.
Comparison of peak-picking workflows for untargeted liquid chromatography/high-resolution mass spectrometry metabolomics data analysis.
Rapid communications in mass spectrometry : RCM (2014-12-03)
Atefeh Rafiei, Lekha Sleno
ABSTRACT
Data analysis is a key step in mass spectrometry based untargeted metabolomics, starting with the generation of generic peak lists from raw liquid chromatography/mass spectrometry (LC/MS) data. Due to the use of various algorithms by different workflows, the results of different peak-picking strategies often differ widely. Raw LC/HRMS data from two types of biological samples (bile and urine), as well as a standard mixture of 84 metabolites, were processed with four peak-picking softwares: Peakview®, Markerview™, MetabolitePilot™ and XCMS Online. The overlaps between the results of each peak-generating method were then investigated. To gauge the relevance of peak lists, a database search using the METLIN online database was performed to determine which features had accurate masses matching known metabolites as well as a secondary filtering based on MS/MS spectral matching. In this study, only a small proportion of all peaks (less than 10%) were common to all four software programs. Comparison of database searching results showed peaks found uniquely by one workflow have less chance of being found in the METLIN metabolomics database and are even less likely to be confirmed by MS/MS. It was shown that the performance of peak-generating workflows has a direct impact on untargeted metabolomics results. As it was demonstrated that the peaks found in more than one peak detection workflow have higher potential to be identified by accurate mass as well as MS/MS spectrum matching, it is suggested to use the overlap of different peak-picking workflows as preliminary peak lists for more rugged statistical analysis in global metabolomics investigations.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodio idrossido, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
Sodio idrossido, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Idrossido di sodio, 50% in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Idrossido di sodio, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Idrossido di sodio, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Sodio idrossido, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodio idrossido, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Acido formico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acido formico, ACS reagent, ≥88%
Sigma-Aldrich
Sodio idrossido, reagent grade, 97%, powder
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodio idrossido, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodio idrossido, pellets, semiconductor grade, 99.99% trace metals basis