Merck
  • Development of analytical method for catechol compounds in mouse urine using hydrophilic interaction liquid chromatography with fluorescence detection.
Development of analytical method for catechol compounds in mouse urine using hydrophilic interaction liquid chromatography with fluorescence detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-16)
Takahiro Kanamori, Muneki Isokawa, Takashi Funatsu, Makoto Tsunoda
ABSTRACT
An analytical method for catecholamines and related compounds using hydrophilic interaction liquid chromatography (HILIC) with native fluorescence detection has been developed. We found that ZIC-cHILIC with phosphorylcholine was suitable for the separation of catechol compounds with good peak shapes among six different HILIC columns (Inertsil SIL, Inertsil Amide, Inertsil Diol, TSKgel NH2-100, ZIC-HILIC, and ZIC-cHILIC). Using ZIC-cHILIC, eight catechol compounds (dopamine, epinephrine, norepinephrine, 3,4-dihydroxyphenylalanine, 3,4-dihydroxyphenylacetic acid, 3,4-dihydroxyphenylglycol, 3,4-dihydroxymandelic acid, and internal standard 3,4-dihydroxybenzylamine) were separated within 15min. The limit of detection at a signal to noise ratio of 3 was 3-28nM. An improved sensitivity was obtained as compared to that of reversed-phase liquid chromatography. This was partly attributed to the increase in the fluorescence intensity of the catechol compounds in the acetonitrile-rich mobile phase. Solid phase extraction using a monolithic silica disk-packed spin column with phenylboronate moieties, which have affinity to catechol compounds, was performed for the selective extraction of catechol compounds from mouse urine. Dopamine, epinephrine, norepinephrine, 3,4-dihydroxyphenylalanine, and 3,4-dihydroxyphenylglycol were successfully quantified in mouse urine.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Fosfato di potassio, ACS reagent, ≥98%
Sigma-Aldrich
Acido formico, reagent grade, ≥95%
Sigma-Aldrich
Acido fosforico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido formico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acido formico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acido fosforico, ACS reagent, ≥85 wt. % in H2O
Supelco
Formato di ammonio, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Acido fosforico, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Formato di ammonio, reagent grade, 97%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Acido acetico, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acido formico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)