Merck
Visualization of integrin Mac-1 in vivo.
Journal of immunological methods (2015-09-08)
Kihong Lim, Young-Min Hyun, Kris Lambert-Emo, David J Topham, Minsoo Kim
ABSTRACT
β2 integrins play critical roles in migration of immune cells and in the interaction with other cells, pathogens, and the extracellular matrix. Among the β2 integrins, Mac-1 (Macrophage antigen-1), composed of CD11b and CD18, is mainly expressed in innate immune cells and plays a major role in cell migration and trafficking. In order to image Mac-1-expressing cells both in live cells and mouse, we generated a knock-in (KI) mouse strain expressing CD11b conjugated with monomeric yellow fluorescent protein (mYFP). Expression of CD11b-mYFP protein was confirmed by Western blot and silver staining of CD11b-immunoprecipitates and total cell lysates from the mouse splenocytes. Mac-1-mediated functions of the KI neutrophils were comparable with those in WT cells. The fluorescence intensity of CD11b-mYFP was sufficient to image CD11b expressing cells in live mice using intravital two-photon microscopy. In vitro, dynamic changes in the intracellular localization of CD11b molecules could be measured by epifluorescent microscopy. Finally, CD11b-expressing immune cells from tissue were easily detected by flow cytometry without anti-CD11b antibody staining.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sodio dodecil solfato, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodio dodecil solfato, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodio dodecil solfato, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodio dodecil solfato, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodio dodecil solfato, BioUltra, 20% in H2O
Supelco
Sodio dodecil solfato, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
N-Iodosuccinimide, 95%
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
Sodio dodecil solfato, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodio dodecil solfato, ≥98.0% (GC)
Sigma-Aldrich
Sodio dodecil solfato, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodio dodecil solfato, ≥90% ((Assay))
Sigma-Aldrich
Sodio dodecil solfato, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodio dodecil solfato, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri