Skip to Content
Merck

GF67039791

Tantalum

foil, 8mm disks, thickness 0.03mm, as rolled, 99.9%

Synonym(s):

Tantalum, TA000348, Ta

Sign In to View Organizational & Contract Pricing.

Select a Size

10 EA

HUF 419,000.00

HUF 419,000.00


Please contact Customer Service for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141741
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tantalum, foil, 8mm disks, thickness 0.03mm, as rolled, 99.9%

InChI

1S/Ta

SMILES string

[Ta]

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

vapor pressure

<0.01 mmHg ( 537.2 °C)

assay

≥99.9%

form

foil

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 670-397-91

resistivity

13.5 μΩ-cm, 20°C

diam. × thickness

8 mm × 0.03 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF27272167GF47620065GF17934914
manufacturer/tradename

Goodfellow 670-397-91

manufacturer/tradename

Goodfellow 272-721-67

manufacturer/tradename

Goodfellow 476-200-65

manufacturer/tradename

Goodfellow 179-349-14

form

foil

form

foil

form

foil

form

foil

assay

≥99.9%

assay

99.90%

assay

99.9%

assay

99.90%

resistivity

13.5 μΩ-cm, 20°C

resistivity

13.5 μΩ-cm, 20°C

resistivity

13.5 μΩ-cm, 20°C

resistivity

13.5 μΩ-cm, 20°C

density

16.69 g/cm3 (lit.)

density

16.69 g/cm3 (lit.)

density

16.69 g/cm3 (lit.)

density

16.69 g/cm3 (lit.)

mp

2996 °C (lit.)

mp

2996 °C (lit.)

mp

2996 °C (lit.)

mp

2996 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of tantalum foil as a mechanical block for the separation of spinal cord and brain in chick embryos.
V HAMBURGER
The Anatomical record, 94, 355-355 (1946-03-01)
Xianfeng Yang et al.
Journal of the American Chemical Society, 131(34), 12048-12049 (2009-08-27)
A rutile microstructure with a novel cross-medal morphology was prepared via a hydrothermal method by using a Tantalum foil as both the doping source and the substrate. This rutile cross-medal prefers to grow along an unprecedented preferential orientation of <110>
Hongbin Yu et al.
PloS one, 8(6), e66447-e66447 (2013-06-27)
We report a simple method to fabricate nano-porous tantalum oxide films via anodization with Ta foils as the anode at room temperature. A mixture of ethylene glycol, phosphoric acid, NH4F and H2O was used as the electrolyte where the nano-porous
Observations on the use of tantalum foil in peripheral nerve surgery.
N C NORCROSS et al.
Journal of neurosurgery, 4(1), 69-71 (1947-01-01)
[The experimental and clinical studies on the anastomosis of the intercostal nerve to the splanchnic nerve by means of metal tantalum foil (an operative procedure on the so-called abdominal neurosis)].
S Bando
Kobe Ika Daigaku kiyo, 26(3), 98-112 (1964-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service