Skip to Content
Merck

BDPH9BCD00A4

Bide Pharmatech

2-Hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid

Synonym(s):

2-hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O6N1P1
CAS Number:
Molecular Weight:
247.14
MDL number:
Product BDPH9BCD00A4 is not currently sold in your country.


Inchi Code

1S/C8H10NO6P/c10-7(11)8(12,16(13,14)15)4-6-2-1-3-9-5-6/h1-3,5,12H,4H2,(H,10,11)(H2,13,14,15)

Inchi Key

FJVYPXVLXQXDHM-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

99%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to off-white solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library