Skip to Content
Merck

HCYTASF-60K

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A Serum-Free

Configurable Serum Matrix-Free Human Cytokine/Chemokine/Growth Factor 48-Plex Panel A

Synonym(s):

human cytokine panel, human growth factor panel

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116131
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A Serum-Free, Configurable Serum Matrix-Free Human Cytokine/Chemokine/Growth Factor 48-Plex Panel A

usage

 kit sufficient for 96 wells

species reactivity

human

packaging

pkg of 1 ea

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

input

cell culture supernatant

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Application

Simultaneously analyze up to 48 analytes in human cell culture supernatants.Analytes included: sCD40L, EGF, Eotaxin/CCL11, FGF-2/FGF-basic, FLT-3L, Fractalkine/CX3CL1, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12(p70), IL-13, IL-15, IL-17A/CTLA8, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10/CXCL10, MCP-1/CCL2, MCP-3/CCL7, M-CSF, MDC, MIG/CXCL9, MIP-1α/CCL3, MIP-1β/CCL4, PDGF-AA, PDGF-AB/BB, RANTES/CCL5, TGFα, TNFα, TNFβ/LTA, VEGF-A

Disclaimer

For research use only. Not for use in diagnostic procedures.Label License/Sticker for Assay Product:By opening the packaging containing this Assay Product (which contains fluorescently labeled microsphere beads authorized by Luminex Corporation) or using this Assay Product in any manner, you are consenting and agreeing to be bound by the End User Terms and Conditions and the End User License Agreement available at http://support.diasorin.com/end-user-terms-and-conditions/. If you do not agree to all of the terms and conditions, you must promptly return this Assay Product for a full refund prior to using it in any manner.

Features and Benefits

48-Plex Power, Simplified: One kit unlocks 48 biomarkers—replaces multiple ELISAs and slashes costs.More Than 20-Year Multiplexing Legacy: The original multiplex innovator—trusted by top pharma companies for critical assays.Cancer, Autoimmunity, & Beyond: Power research in immunotherapy, sepsis, Alzheimer′s, and Long COVID biomarker discovery.Intuitive Workflow: User-friendly protocols that require minimal training, allowing researchers to achieve publication-ready results without extensive background in multiplex assays.Configurable Assay Design: Configure your panel by selecting specific analytes based on your research needs, allowing for a tailored approach that maximizes relevance and impact.Peer-Reviewed Credibility: Frequently cited in immunology and clinical research publications. Trusted by academic institutions, CROs, and pharma companies.Designed for Human Cell Culture Supernatants: Optimized for analyzing cytokines, chemokines, and growth factors in cell culture supernatants.

General description

Cytokines, chemokines, and growth factors are key mediatorsof immune system functions capable of signaling through autocrine, paracrine,and endocrine mechanisms. Their pleiotropic immunomodulatory propertiesallow these biomolecules to react to diverse stimuli and regulate the immuneresponse, either promoting or inhibiting inflammation.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

target_organs

Respiratory Tract

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service