Skip to Content
Merck

95146

Retinol solution

≥95.0% (HPLC)

Synonym(s):

3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexenyl)-2,4,6,8-nonatetraen-1-ol, Axerophthol, Vitamin A alcohol solution, Vitamin A1, all-trans-Retinol

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C20H30O
CAS Number:
Molecular Weight:
286.45
Beilstein:
403040
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058018
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

human plasma

Assay

≥95.0% (HPLC)

form

liquid

color

colorless to very light yellow

mp

62-64°C (solvent free)

storage temp.

2-8°C

SMILES string

CC(=C/CO)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H30O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,21H,7,10,14-15H2,1-5H3/b9-6+,12-11+,16-8+,17-13+

InChI key

FPIPGXGPPPQFEQ-OVSJKPMPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain.
Retinol is biological active in a wide range of processes.

Analysis Note

0.25 mg in 1 mL hexane

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Spencer et al.
Science (New York, N.Y.), 343(6169), 432-437 (2014-01-25)
How the immune system adapts to malnutrition to sustain immunity at barrier surfaces, such as the intestine, remains unclear. Vitamin A deficiency is one of the most common micronutrient deficiencies and is associated with profound defects in adaptive immunity. Here
Jacqueline A Jumpsen et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 5(2), 77-84 (2006-03-02)
The objective of this study was to investigate the effect of docosahexaenoic acid (DHA) supplementation on blood and intestinal DHA levels and lung function in mild/moderately affected adult CF patients with the DeltaF508 genotype. Cystic Fibrosis (CF) patients often present
K R Norum et al.
The American journal of clinical nutrition, 56(4), 735-744 (1992-10-01)
We discuss vitamin A with emphasis on its absorption, transport, cellular uptake, storage, and intracellular metabolism. Dietary retinyl esters are hydrolyzed to retinol in the intestinal lumen before absorption by enterocytes. Carotenoids are absorbed and then partially converted to retinol
N M Hamdan et al.
Journal of dental research, 92(12 Suppl), 146S-153S (2013-10-26)
People wearing mandibular two-implant overdentures (IOD) chew food with less difficulty than those wearing conventional complete dentures (CD). However, there is still controversy over whether or not this results in better dietary intake. In this randomized clinical trials (RCT), the
Frank T Wieringa et al.
The American journal of clinical nutrition, 77(3), 651-657 (2003-02-26)
Deficiencies of iron and vitamin A are prevalent worldwide. Single-micronutrient supplementation is widely used to combat these deficiencies. However, micronutrient deficiencies often occur concurrently, and there are many interactions between micronutrients. This study investigated interactions among 3 important micronutrients--iron, vitamin

Related Content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service