1724736
USP
Zinc Chloride
United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
Select a Size
100 MG
HUF 219,000.00
HUF 219,000.00
Please contact Customer Service for Availability
About This Item
Empirical Formula (Hill Notation):
Cl2Zn
CAS Number:
Molecular Weight:
136.30
UNSPSC Code:
41116107
Skip To
grade
pharmaceutical primary standard
Agency
USP
form
crystalline
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service