Merck
  • Influence of A-21T and C-262T genetic polymorphisms at the promoter region of the catalase (CAT) on gene expression.
Influence of A-21T and C-262T genetic polymorphisms at the promoter region of the catalase (CAT) on gene expression.
Environmental Health and Preventive Medicine (2016)
Saify K, et al.