- Enhanced p62 expression triggers concomitant autophagy and apoptosis in a rat chronic spinal cord compression model.
Enhanced p62 expression triggers concomitant autophagy and apoptosis in a rat chronic spinal cord compression model.
Molecular Medicine Reports (2014)
Chen Z, et al.