Merck
  • Sixty years from discovery to solution: crystal structure of bovine liver catalase form III
Sixty years from discovery to solution: crystal structure of bovine liver catalase form III
Acta Crystallographica Section D, Biological Crystallography (2011)
Foroughi LM, et al.