Merck
  • Synthetic Peptide Purification via Solid-Phase Extraction with Gradient Elution: A Simple, Economical, Fast, and Efficient Methodology.
Synthetic Peptide Purification via Solid-Phase Extraction with Gradient Elution: A Simple, Economical, Fast, and Efficient Methodology.
Molecules (Basel, Switzerland) (2019-03-31)
Diego Sebastián Insuasty Cepeda, Héctor Manuel Pineda Castañeda, Andrea Verónica Rodríguez Mayor, Javier Eduardo García Castañeda, Mauricio Maldonado Villamil, Ricardo Fierro Medina, Zuly Jenny Rivera Monroy
ABSTRACT
A methodology was implemented for purifying peptides in one chromatographic run via solid-phase extraction (SPE), reverse phase mode (RP), and gradient elution, obtaining high-purity products with good yields. Crude peptides were analyzed by reverse phase high performance liquid chromatography and a new mathematical model based on its retention time was developed in order to predict the percentage of organic modifier in which the peptide will elute in RP-SPE. This information was used for designing the elution program of each molecule. It was possible to purify peptides with different physicochemical properties, showing that this method is versatile and requires low solvent consumption, making it the least polluting one. Reverse phase-SPE can easily be routinely implemented. It is an alternative to enrich and purified synthetic or natural molecules.