Merck
  • N-docosahexaenoylethanolamine reduces neuroinflammation and cognitive impairment after mild traumatic brain injury in rats.
N-docosahexaenoylethanolamine reduces neuroinflammation and cognitive impairment after mild traumatic brain injury in rats.
Scientific reports (2021-01-14)
Arina I Ponomarenko, Anna A Tyrtyshnaia, Evgeny A Pislyagin, Inessa V Dyuizen, Ruslan M Sultanov, Igor V Manzhulo
ABSTRACT
At present, there is a growing interest in the study of the neurotropic activity of polyunsaturated fatty acids ethanolamides (N-acylethanolamines). N-docosahexaenoylethanolamine (DHEA, synaptamide) is an endogenous metabolite and structural analogue of anandamide, a widely studied endocannabinoid derived from arachidonic acid. The results of this study demonstrate that DHEA, when administered subcutaneously (10 mg/kg/day, 7 days), promotes cognitive recovery in rats subjected to mild traumatic brain injury (mTBI). In the cerebral cortex of experimental animals, we analyzed the dynamics of Iba-1-positive microglia activity changes and the expression of pro-inflammatory markers (IL1β, IL6, CD86). We used immortalized mouse microglial cells (SIM-A9) to assess the effects of DHEA on LPS-induced cytokines/ROS/NO/nitrite, as well as on CD206 (anti-inflammatory microglia) and the antioxidant enzyme superoxide dismutase (SOD) production. In vivo and in vitro experiments showed that DHEA: (1) improves indicators of anxiety and long-term memory; (2) inhibits the pro-inflammatory microglial cells activity; (3) decrease the level of pro-inflammatory cytokines/ROS/NO/nitrites; (4) increase CD206 and SOD production. In general, the results of this study indicate that DHEA has a complex effect on the neuroinflammation processes, which indicates its high therapeutic potential.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Rat CD86 / CD86 Antigen, Isoform CRA-c ELISA Kit
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests