Merck
  • Revisiting the isolation and characterisation of Entamoeba histolytica lipopeptidophosphoglycan.
Revisiting the isolation and characterisation of Entamoeba histolytica lipopeptidophosphoglycan.
Parasitology research (2024-02-21)
Anna Nagode, Jorick Vanbeselaere, Michael Duchêne
ABSTRACT
The parasite Entamoeba histolytica is the cause of amoebic dysentery and liver abscess in humans. On the protozoan cell surface, a variety of glycosylated molecules are involved in the interaction with the environment, such as attachment to the colonic mucus. One of these molecules is the lipopeptidophosphoglycan (LPPG), a complex surface component with antigenic properties. Its structure is only partly known, it is a glycosylphosphatidylinositol (GPI)-linked glycoprotein with a large amount of O-glycosylation. To date, the sequence of a core protein has not been identified. In this study, we further investigated this complex surface molecule aided by the availability of the monoclonal antibody EH5, which had been raised in our laboratory. We studied the extraction of LPPG in various solvent mixtures and discovered that 2-butanol saturated water was simple and superior to other solvents used in the past. The isolated LPPG was subjected to treatment with several proteases and the Ser/Thr specific cleavage agent scandium (III) trifluoromethanesulfonate (scandium triflate). The products were probed with antibody EH5 and the blots showed that the LPPG preparation was largely resistant to standard proteases, but could be cleaved by the scandium compound. These observations could point to the existence of a Ser- or Thr-rich core protein structure.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution