Merck
  • Sodium dodecyl sulphate-polyacrylamide gel electrophoresis of human parotid salivary proteins: comparison of dansylation, coomassie blue R-250 and silver detection methods.
Sodium dodecyl sulphate-polyacrylamide gel electrophoresis of human parotid salivary proteins: comparison of dansylation, coomassie blue R-250 and silver detection methods.
Electrophoresis (1996-03-01)
J A Beeley, F Newman, P H Wilson, I C Shimmin
ABSTRACT
Detection of human parotid salivary proteins by dansylation and UV-transillumination after sodium dodecyl sulphate-polyacrylamide gel electrophoresis (SDS-PAGE) has been compared with Coomassie Blue R-250 and silver staining procedures. Dansylation gives superior results in terms of both resolution and sensitivity, especially with basic proline-rich proteins (PRPs).
Sigma-Aldrich
Brilliant Blue R, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue R, pure
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade