Merck
  • Encapsulation and enzyme-mediated release of molecular cargo in polysulfide nanoparticles.
Encapsulation and enzyme-mediated release of molecular cargo in polysulfide nanoparticles.
ACS nano (2011-05-21)
Brett L Allen, Jermaine D Johnson, Jeremy P Walker
ABSTRACT
Poly(propylene sulfide) nanoparticles (<150 nm) have been synthesized by an anionic, ring-opening emulsion polymerization. Upon exposure to parts per million (ppm) levels of oxidizing agent (NaOCl), hydrophobic polysulfide particles are oxidized to hydrophilic polysulfoxides and polysulfones. Utilizing this mechanism, the encapsulation of hydrophobic molecular cargo, including Nile red and Reichardt's dye, within polysulfide nanoparticles has been characterized by a variety of microscopic and spectroscopic methods and its release demonstrated via chemical oxidation. Moreover, release of cargo has been enzymatically driven by oxidoreductase enzymes such as chloroperoxidase and myeloperoxidase in the presence of low concentrations of sodium chloride (200 mM) and hydrogen peroxide (500 μM). This oxidation-driven mechanism holds promise for controlled encapsulation and release of a variety of hydrophobic cargos.
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, aqueous suspension, brown, >10,000 U/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, ≥3,000 units/mL