Merck
  • Experimental and theoretical studies on formation and degradation of chloro organic compounds.
Experimental and theoretical studies on formation and degradation of chloro organic compounds.
Chemosphere (2005-09-09)
Piotr Cysewski, Alicja Gackowska, Jerzy Gaca
ABSTRACT
Studies on oxidation of tert-butyl ethers in the presence of chloride ions proved that acid medium favoured formation of chloro organic compounds. 1,2-Dichloro-2-methylpropane, 3-chloro-2-chloromethylpropene were identified among the reaction products. Presence of these compounds was identified both in the case when methyl-tert-butyl ether (MTBE) and ethyl-tert-butyl ether (ETBE) were subjected to reaction. Reaction products were analysed by gas chromatography method with application of -FID, -MS and -AED detectors. On the basis of experimental data, the path of tert-butyl ethers conversion to dichloro products was proposed. It was found that the identified chloro derivatives could be formed both by ionic and radical reactions. In order to confirm this thesis for the proposed scheme of reaction, the theoretical calculations of molecular simulation of the reaction paths were performed.
Product Description
Supelco
1,2-Dichloropropane, analytical standard
Sigma-Aldrich
1,2-Dichloropropane, 99%
Sigma-Aldrich
3-Chloro-2-methyl-1-propene, contains ~250 ppm Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) as stabilizer, 98%
Sigma-Aldrich
3-Chloro-2-methyl-1-propene, technical grade, 90%