Merck
  • Sequential Cu-catalyzed amidation-base-mediated camps cyclization: a two-step synthesis of 2-Aryl-4-quinolones from o-halophenones.
Sequential Cu-catalyzed amidation-base-mediated camps cyclization: a two-step synthesis of 2-Aryl-4-quinolones from o-halophenones.
The Journal of organic chemistry (2007-09-14)
Carrie P Jones, Kevin W Anderson, Stephen L Buchwald
ABSTRACT
A direct two-step method for the preparation of 2-aryl- and 2-vinyl-4-quinolones that utilizes a copper-catalyzed amidation of o-halophenones followed by a base-promoted Camps cyclization of the resulting N-(2-ketoaryl)amides is described. With CuI, a diamine ligand, and base as the catalyst system, the amidation reactions proceed in good yields for a range of aryl, heteroaryl, and vinyl amides. The subsequent Camps cyclization efficiently provides the desired 4-quinolones with the conditions that are described.
Product Description
Supelco
2-Bromoacetophenone, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
2-Bromoacetophenone, 98%