Merck
  • Expression of renewal is dependent on the extinction-test interval rather than the acquisition-extinction interval.
Expression of renewal is dependent on the extinction-test interval rather than the acquisition-extinction interval.
Behavioral neuroscience (2009-06-03)
Jee Hyun Kim, Rick Richardson
ABSTRACT
A recent finding suggested that when extinction occurs shortly after acquisition, renewal of an extinguished fear response (fear-potentiated startle) to a light conditioned stimulus (CS) is diminished (Myers, Ressler, & Davis, 2006). The present study attempted to extend this finding using a white-noise CS and freezing as the behavioral measure of fear. In Experiments 1A and 1B, we observed renewal whether extinction occurred 10 min or 24 hr after acquisition. In contrast, renewal was not observed if test occurred 10 min after extinction (Experiment 2). Experiment 3 demonstrated that expression of extinction at the 10-min extinction-test interval was attenuated by a pretest subcutaneous injection of the gamma-aminobutyric acid (GABA) inverse agonist FG7142. These findings suggest that renewal is influenced more by the extinction-test interval than the acquisition-extinction interval. Further, the failure to see renewal 10 min after extinction suggests that there is a separate context memory that undergoes a different consolidation function than the CS-no US memory formed during extinction. Finally, the expression of extinction appears to be GABA dependent regardless of the extinction-test interval or the test context.
Sigma-Aldrich
β-Carboline-3-carboxylic acid N-methylamide
Pricing and availability is not currently available.