- Purification of lysyl oxidase from piglet skin by selective interaction with Sephacryl S-200.
Purification of lysyl oxidase from piglet skin by selective interaction with Sephacryl S-200.
The Biochemical journal (1990-03-15)
D R Shackleton, D J Hulmes
PMID1970238
ABSTRACT
A method is described for the purification of piglet skin lysyl oxidase from a crude urea extract by two simple chromatographic procedures. The method relies on the selective interaction between Sephacryl S-200 and lysyl oxidase that occurs in the absence of urea and in conditions of low ionic strength.