Merck
  • Template induced structural isomerism and enhancement of porosity in manganese(II) based metal-organic frameworks (Mn-MOFs).
Template induced structural isomerism and enhancement of porosity in manganese(II) based metal-organic frameworks (Mn-MOFs).
Chemical communications (Cambridge, England) (2011-06-11)
Tamas Panda, Pradip Pachfule, Rahul Banerjee
ABSTRACT
Three new metal-organic framework isomers have been synthesized by using the organic linker 5-triazole isophthalic acid and Mn(NO(3))(2)·xH(2)O. Structural conversions from non-porous to porous MOFs due to the template effect have been observed. The cross-sectional pore apertures of the resulting Mn-MOFs are comparable to the molecular dimensions of the template (pyrazine and 4,4'-bipyridine). The periodic increased porosity in Mn-MOFs depending on the size of the template used has been further confirmed by the CO(2) adsorption isotherms.
Sigma-Aldrich
4,4′−Dipyridyl, 98%
Sigma-Aldrich
Isophthalic acid, 99%