Merck
  • Fluorescent studies of salicylaldehyde and other related carbonyl compounds for the selective and sensitive detection of zinc(II) ions in aqueous solution.
Fluorescent studies of salicylaldehyde and other related carbonyl compounds for the selective and sensitive detection of zinc(II) ions in aqueous solution.
Journal of fluorescence (2011-11-05)
Soleiman Hisaindee, Osama Zahid, Mohammed A Meetani, John Graham
ABSTRACT
Salicylaldehyde was found to have a high selectivity for zinc ions with simultaneous enhancement of fluorescence in aqueous buffer solution at optimum pH 8.5. The stoichiometry of the complex was determined to be 1:1 with a K(a) value of 3.4 × 10(4) M(-1) at 298 K. The fluorescence of the complex is not affected by common anions and Zn(2+) binds preferentially to salicylaldehyde in the presence of alkali, alkaline earth and heavy metal cations (Hg(2+), Cd(2+), Cr(3+) and Ni(2+)). This property is not observed with related phenolic compounds bearing a carbonyl group such as esters, amides, carboxylic acids and ketones.
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%