Merck
  • Lateral septal area α1- and α2-adrenoceptors differently modulate baroreflex activity in unanaesthetized rats.
Lateral septal area α1- and α2-adrenoceptors differently modulate baroreflex activity in unanaesthetized rats.
Experimental physiology (2011-12-20)
América A Scopinho, Fernando H F Alves, Leonardo B M Resstel, Fernando M A Correa, Carlos C Crestani
ABSTRACT
The lateral septal area (LSA) is a limbic structure involved in autonomic, neuroendocrine and behavioural responses. An inhibitory influence of the LSA on baroreflex activity has been reported; however, the local neurotransmitter involved in this modulation is still unclear. In the present study, we verified the involvement of local LSA adrenoceptors in modulating cardiac baroreflex activity in unanaesthetized rats. Bilateral microinjection of the selective α(1)-adrenoceptor antagonist WB4101 (10 nmol in a volume of 100 nl) into the LSA decreased baroreflex bradycardia evoked by blood pressure increases, but had no effect on reflex tachycardia evoked by blood pressure decreases. Nevertheless, bilateral administration of the selective α(2)-adrenoceptor antagonist RX821002 (10 nmol in 100 nl) increased baroreflex tachycardia without affecting reflex bradycardia. Treatment of the LSA with a cocktail containing WB4101 and RX821002 decreased baroreflex bradycardia and increased reflex tachycardia. The non-selective β-adrenoceptor antagonist propranolol (10 nmol in 100 nl) did not affect either reflex bradycardia or tachycardia. Microinjection of noradrenaline into the LSA increased reflex bradycardia and decreased the baroreflex tachycardic response, an opposite effect compared with those observed after double blockade of α(1)- and α(2)-adrenoceptors, and this effect of noradrenaline was blocked by local LSA pretreatment with the cocktail containing WB4101 and RX821002. The present results provide advances in our understanding of the baroreflex neural circuitry. Taken together, data suggest that local LSA α(1)- and α(2)-adrenoceptors modulate baroreflex control of heart rate differently. Data indicate that LSA α(1)-adrenoceptors exert a facilitatory modulation on baroreflex bradycardia, whereas local α(2)-adrenoceptors exert an inhibitory modulation on reflex tachycardia.
Product Description
Supelco
WB-4101 hydrochloride, analytical standard
Sigma-Aldrich
Idazoxan hydrochloride
Sigma-Aldrich
RX 821002 hydrochloride, solid, ≥98% (HPLC)