Merck
Hyaladione, an S-methyl cyclohexadiene-dione from Hyalangium minutum.
Journal of natural products (2012-04-14)
Patrick W Okanya, Kathrin I Mohr, Klaus Gerth, Heinrich Steinmetz, Volker Huch, Rolf Jansen, Rolf Müller
ABSTRACT
A bioassay-guided fractionation of the crude methanol extract of the myxobacterium Hyalangium minutum, strain NOCB-2(T) (DSM 14724(T)), led to the isolation of hyaladione (1), a novel S-methyl cyclohexadiene-dione. The structure of 1 was established by HRESIMS, NMR, and IR spectroscopy as well as X-ray crystallography. Compound 1 was active against growing mammalian cell lines, with IC(50) values ranging from 1.23 to 3.93 μM, in addition to a broad spectrum of antibacterial and antifungal activities, including inhibition of pathogenic methicillin-resistant Staphylococcus aureus and Pseudomonas aeruginosa with an MIC of 0.83 and 8.5 μg mL(-1), respectively.
Sigma-Aldrich
1,4-Cyclohexadiene, 97%
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)