Merck
  • Structure and biosynthesis of fimsbactins A-F, siderophores from Acinetobacter baumannii and Acinetobacter baylyi.
Structure and biosynthesis of fimsbactins A-F, siderophores from Acinetobacter baumannii and Acinetobacter baylyi.
Chembiochem : a European journal of chemical biology (2013-03-05)
Anna Proschak, Patrice Lubuta, Peter Grün, Frank Löhr, Gottfried Wilharm, Veronique De Berardinis, Helge B Bode
ABSTRACT
Novel chatechol/hydroxamate siderophores (named "fimsbactins") were identified in Acinetobacter baumannii ATCC 17978 and Acinetobacter baylyi ADP1. The major compound, fimsbactin A, was isolated from low-iron cultures of A. baylyi ADP1, and its chemical structure was elucidated by mass spectrometry, and detailed (1)H, (13)C and (15)N NMR spectroscopy. From inverse feeding experiments following HPLC-MS analysis, the structures of five additional derivatives were elucidated. The gene cluster encoding the fimsbactin synthetase (fbs) was identified in both genomes, and mutants in fbs genes in A. baylyi were analyzed, thus allowing prediction of the fimsbactin biosynthesis pathway.
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%